missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human HVEM/TNFRSF14 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems MAB3563-100
This item is not returnable.
View return policy
Description
HVEM/TNFRSF14 Monoclonal antibody specifically detects HVEM/TNFRSF14 in Human samples. It is validated for Flow Cytometry, Neutralization
Specifications
| HVEM/TNFRSF14 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| ATAR, CD270, CD270 antigen, CD40-like protein, Herpes virus entry mediator A, Herpesvirus entry mediator A, HveA, HVEMTR2HVEAATAR, LIGHTR, TNFRSF14, tumor necrosis factor receptor superfamily member 14, tumor necrosis factor receptor superfamily, member 14 (herpesvirus entrymediator), Tumor necrosis factor receptor-like 2, tumor necrosis factor receptor-like gene2 | |
| Human embryonic kidney cell HEK293-derived human HVEM/TNFRSF14, Pro37-Val202, Accession # Q92956 | |
| 100 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
| IgG |
| Flow Cytometry, Neutralization | |
| 2742B | |
| Flow Cytometry 0.25 ug/10^6 cells, Neutralization | |
| Q92956 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 8764 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction