missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IFN-alpha/beta R2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
206.00 € - 461.00 €
Specifications
| Antigen | IFN-alpha/beta R2 |
|---|---|
| Clone | 493715 |
| Dilution | Neutralization 4-20 μg/mL |
| Applications | Neutralization |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18732117
![]() |
R&D Systems
MAB4015-SP |
25 μg |
206.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18734177
|
R&D Systems
MAB4015-100 |
100 μg |
461.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IFN-alpha/beta R2 Monoclonal specifically detects IFN-alpha/beta R2 in Human samples. It is validated for Neutralization.Specifications
| IFN-alpha/beta R2 | |
| Neutralization 4-20 μg/mL | |
| Monoclonal | |
| Supernatant | |
| Human | |
| P48551 | |
| 3455 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
| 493715 | |
| Neutralization | |
| Unconjugated | |
| Mouse | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| beta receptor10, human interferon alpha, human interferon alpha/beta receptor, IFNABR, IFN-alpha binding protein, IFN-alpha/beta receptor 2, IFN-alpha-REC, IFN-aR2, IFNAR2, IFNARB, IFNbR2, IFN-bR2, IFN-R, IFN-R-2, interferon (alpha, beta and omega) receptor 2, Interferon alpha binding protein, interferon alpha/beta receptor 2, interferon-alpha/beta receptor beta chain, Type I interferon receptor 2 | |
| Mouse myeloma cell line, NS0-derived recombinant human IFN-alpha/beta R2, Ile27-Lys243, Accession # P48551 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human IFN-alpha/beta R2 in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title