missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IFN-gamma PerCP-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
288.00 € - 521.00 €
Specifications
| Antigen | IFN-gamma |
|---|---|
| Clone | 25723 |
| Dilution | Intracellular Staining by Flow Cytometry 10 uL/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18702579
![]() |
R&D Systems
IC285C-025 |
25 Tests |
288.00 €
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18702667
![]() |
R&D Systems
IC285C-100 |
100 Tests |
521.00 €
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IFN-gamma Monoclonal specifically detects IFN-gamma in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
| IFN-gamma | |
| Intracellular Staining by Flow Cytometry 10 uL/10^6 cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| IFG, IFI, IFNG, IFNgamma, IFN-gamma, Immune interferon, interferon gamma, interferon, gamma | |
| IFNG | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Detects human IFN-gamma in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse IFN-gamma , recombinant rat IFN-gamma , or recombinant porcine IFN-gamma is observed. |
| 25723 | |
| Flow Cytometry | |
| PerCP | |
| Mouse | |
| Human | |
| AAP20098.1 | |
| 3458 | |
| E. coli-derived recombinant human IFN-gamma Gln24-Gln166 Accession # AAP20098.1 | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts