Learn More
Human IFN-kappa Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  R&D Systems MAB102472-100
 View more versions of this product
Product Code. 18781477
  • 423.00 € / 100µg
Description
IFN-kappa Monoclonal specifically detects IFN-kappa in Human samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IFN-kappa
Monoclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
IFNK, IFNkappa, IFN-kappa
E. coli-derived recombinant human IFN-kappa , Leu28-Lys207, Accession # Q90W0
100 μg
56832
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
IgG2a
ELISA
1009727
ELISA
Q90W0
Mouse
Protein A or G purified from hybridoma culture supernatant
Primary
Detects human IFN-kappa in direct ELISAs.
Human
Lyophilized