missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IGF-I R/IGF1R Fluorescein-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems FAB391F-100
This item is not returnable.
View return policy
Description
IGF-I R/IGF1R Monoclonal specifically detects IGF-I R/IGF1R in Human samples. It is validated for Flow Cytometry.
Specifications
| IGF-I R | |
| Monoclonal | |
| Fluorescein | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| CD221, CD221 antigen, EC 2.7.10, EC 2.7.10.1, IGF1R, IGF-1R, IGF-I receptor, IGFIR, IGF-IR, IGFR, insulin-like growth factor 1 receptor, Insulin-like growth factor I receptor, JTK13, MGC142170, MGC142172, MGC18216, soluble IGF1R variant 1, soluble IGF1R variant 2 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3480 | |
| Human | |
| Purified |
| Flow Cytometry | |
| 33255 | |
| Flow Cytometry 10 uL/10^6 cells | |
| P08069 | |
| IGF1R | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IGF-I R Glu31-Asn932 Accession # P08069 | |
| 100 Tests | |
| Primary | |
| Detects human IGF-I R in sandwich immunoassays and Western blots. In sandwich immunoassays, less than 0.15% cross-reactivity or interference was observed with recombinant human (rh) IGF-I, rhIGF-II, rhIL-3 R alpha , rhIL-9 R, and rhTGF-beta RII. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction