Learn More
Human IgG Fc Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody has been used in 4 publications
Brand:  R&D Systems MAB110-500
 View more versions of this product
Product Code. 18758408
  • 2100.00 € / 500µg
Description
IgG Monoclonal specifically detects IgG in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IgG
Monoclonal
LYOPH
Western Blot 1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready
Immunoglobulin G, ImmunoglobulinG
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
3500
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG1
Western Blot, Flow Cytometry, CyTOF
97924
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS and NaCl with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
IGHG1
Mouse myeloma cell line NS0-derived recombinant human IgG 1 Fc region
500 μg
Primary
Detects chimeric proteins containing a human IgG Fc region in direct ELISAs and Western blots.
Human
Supernatant