missing translation for 'onlineSavingsMsg'
Learn More

Human IgG2 Antibody, R&D Systems™
GREENER_CHOICE

Rabbit Monoclonal Antibody

214.00 € - 506.00 €

Specifications

Antigen Human IgG2
Clone 2348B
Dilution Western Blot 2 μg;/mL, ELISA
Applications Western Blot, ELISA
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18794507
GREENER_CHOICE
View Documents
R&D Systems
MAB97941-SP
25 μg
214.00 €
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18793237
GREENER_CHOICE
View Documents
R&D Systems
MAB97941-100
100 μg
506.00 €
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Human IgG2 Monoclonal specifically detects Human IgG2 in Human samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

Human IgG2
Western Blot 2 μg;/mL, ELISA
Monoclonal
Supernatant
Human
Ig gamma-2 chain C region, IGHG2, immunoglobulin heavy constant gamma 2 (G2m marker)
Human IgG2 protein
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects human IgG2 in direct ELISAs. It does not detect human IgG1, IgG3 and IgG4 in direct ELISAs and Western Blots.
2348B
Western Blot, ELISA
Unconjugated
Rabbit
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
3501.0
IgG
Protein A or G purified from cell culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.