missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IgM Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems MAB9435-100
This item is not returnable.
View return policy
Description
IgM Monoclonal specifically detects IgM in Human samples. It is validated for Western Blot, Simple Western, ELISA.
Specifications
| IgM | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 3507 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG |
| Western Blot, ELISA | |
| 2123A | |
| Western Blot 0.1 ug/mL, Simple Western 5 ug/mL, ELISA | |
| AGM1, DKFZp686I15196, DKFZp686I15212, FLJ00385, immunoglobulin heavy constant mu, MGC104996, MGC52291, MU, VH | |
| Immunoglobulin M (IgM), mu chain from human plasma | |
| 100 μg | |
| Primary | |
| Detects human IgM in Western blots. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction