missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-1 alpha/IL-1F1 (Research Grade Bermekimab Biosimilar) Antibody, R&D Systems™
Description
IL-1 alpha/IL-1F1 Monoclonal antibody specifically detects IL-1 alpha/IL-1F1 in Human samples. It is validated for Flow Cytometry
Specifications
Specifications
| Antigen | IL-1 alpha/IL-1F1 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | Hu146 |
| Conjugate | Unconjugated |
| Dilution | Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. |
| Gene Alias | BAF, Hematopoietin-1, IL1, IL1 alpha, IL-1 alpha, IL1A, IL-1A, IL1-ALPHA, IL1F1, IL-1F1, IL1F1hematopoietin-1, interleukin 1, alpha, interleukin-1 alpha, LAF, LEM, preinterleukin 1 alpha, pro-interleukin-1-alpha |
| Host Species | Human |
| Immunogen | Human IL-1 alpha/IL-1F1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?