missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-1 beta/IL-1F2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB601R-100
This item is not returnable.
View return policy
Description
IL-1 beta/IL-1F2 Monoclonal specifically detects IL-1 beta/IL-1F2 in Human samples. It is validated for Western Blot, Simple Western, Neutralization, Immunocytochemistry, ELISA Capture (Matched Antibody Pair).
Specifications
| IL-1 beta/IL-1F2 | |
| Monoclonal | |
| Unconjugated | |
| Supplied as a solution in PBS. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| Mouse | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 3553 | |
| Human | |
| IgG1 |
| Western Blot, Neutralization, Immunocytochemistry, ELISA | |
| 2805R | |
| Western Blot 1 ug/mL, Simple Western 10 ug/mL, Neutralization 50-200 ng/mL, Immunocytochemistry 8-25 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| catabolin, IL-1, IL1 beta, IL-1 beta, IL-1b, IL1B, IL1-BETA, IL-1F2, IL1F2IL-1 beta, interleukin 1, beta, interleukin-1 beta, preinterleukin 1 beta, pro-interleukin-1-beta | |
| E. coli-derived recombinant human IL-1 beta/IL-1F2 | |
| 100 μg | |
| Primary | |
| Detects human IL-1 beta/IL-1F2 in sandwich ELISAs and Western blots. In sandwich ELISAs, less than 4% cross-reactivity with recombinant rat (rr) IL-1 beta and less than 0.1% with recombinant porcine (rp) IL-1 beta , recombinant human IL-1 alpha , rpIL-1 alpha , rrIL-1 alpha , recombinant mouse (rm) IL-1 alpha , and rmIL-1 beta is observed. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction