missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-1 RII Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB663-500
This item is not returnable.
View return policy
Description
IL-1 RII Monoclonal specifically detects IL-1 RII in Human samples. It is validated for Western Blot, Neutralization, ELISA Capture (Matched Antibody Pair).
Specifications
| IL-1 RII | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| beta, CD121b, CD121b antigen, IL-1 R beta, IL1R2, IL1RBCD121 antigen-like family member B, IL-1R-beta, IL1RII, IL-1RII, IL-1RT2, IL-1RT-2, interleukin 1 receptor, type II, Interleukin-1 receptor beta, MGC47725 | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 7850 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot, Neutralization, ELISA | |
| 34141 | |
| Western Blot 1 ug/mL, Neutralization 5-20 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| P27930 | |
| IL1R2 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IL-1 RII Phe14-Glu343 (Ser56Gly and Glu297Gly) Accession # P27930 | |
| 500 μg | |
| Primary | |
| Detects human IL-1 RII in direct ELSAs and Western blots. When used in combination with the biotinylated human IL-1 RII affinity purified polyclonal detection antibody (Catalog # BAF263) in sandwich ELISAs, no significant cross-reactivity or interference was observed with recombinant human (rh) IL-1ra, rhIL-1 RI, recombinant mouse IL-1ra, or recombinant rat IL-1ra. | |
| Human | |
| Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction