missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-12 p70 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 19 publications
Brand: R&D Systems MAB219-500
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
IL-12 p70/IL-12A Monoclonal specifically detects IL-12 p70/IL-12A in Human samples. It is validated for Western Blot, Immunohistochemistry, Neutralization.
Specifikationer
| IL-12 p70/IL-12A | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| IL12A | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IL-12 p70 | |
| 500 μg | |
| Primary | |
| Detects human IL-12 p70 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) IL-12 p40 homodimer or rhIL-23 is observed. | |
| Human | |
| Supernatant |
| Western Blot, Immunohistochemistry, Neutralization | |
| 24910 | |
| Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL, Neutralization 0.3-0.9 ug/mL | |
| CLMF p35, CLMF1, Cytotoxic lymphocyte maturation factor 35 kDa subunit, IL12, IL12 p70, IL-12 subunit p35, IL-12, subunit p35, IL12A, interleukin 12, p35, interleukin 12A (natural killer cell stimulatory factor 1, cytotoxic lymphocytematuration factor 1, p35), interleukin-12 alpha chain, interleukin-12 subunit alpha, natural killer cell stimulatory factor 1, 35 kD subunit, NF cell stimulatory factor chain 1, NK cell stimulatory factor chain 1, NKSF1, p35 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3592 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
For Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering