missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-12 R beta 1 Fluorescein-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 1 publication
Brand: R&D Systems FAB839F-025
This item is not returnable.
View return policy
Description
IL-12 R beta 1 Monoclonal specifically detects IL-12 R beta 1 in Human samples. It is validated for Flow Cytometry.
Specifications
| IL-12 R beta 1 | |
| Monoclonal | |
| Fluorescein | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| CD212, CD212 antigen, IL-12 receptor beta component, IL-12 receptor subunit beta-1, IL12R, IL12R beta 1, IL-12R subunit beta-1, IL12RB, IL12RB1, IL-12RB1, IL-12R-beta-1, IL-12R-BETA1, interleukin 12 receptor, beta 1, interleukin-12 receptor beta-1 chain, interleukin-12 receptor subunit beta-1, MGC34454 | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 3594 | |
| Human | |
| Purified |
| Flow Cytometry | |
| 69310 | |
| Flow Cytometry 10 uL/10^6 cells | |
| P42701 | |
| IL12RB1 | |
| Mouse myeloma cell line NS0-derived recombinant human IL-12 R beta 1 Cys24-Glu540 Accession # P42701 | |
| 25 Tests | |
| Primary | |
| Detects human IL-12 R beta 1 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse IL-12 R beta 1 is observed. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction