missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-15R alpha Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems MAB10900-SP
This item is not returnable.
View return policy
Description
IL-15R alpha Monoclonal antibody specifically detects IL-15R alpha in Human samples. It is validated for Flow Cytometry
Specifications
| IL-15R alpha | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. | |
| CD215, CD215 antigen, IL15 R alpha, IL-15 R alpha, IL-15 receptor subunit alpha, IL15RA, IL-15RA, IL-15R-alpha, interleukin 15 receptor, alpha, interleukin-15 receptor subunit alpha, MGC104179 | |
| Mouse myeloma cell line, NS0-derived human IL-15R alpha, Met1-Lys173, Accession # NP_751951 | |
| 25 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
| IgG |
| Flow Cytometry | |
| 2639B | |
| Flow Cytometry 0.25 ug/10^6 cells | |
| NP_751951 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 3601 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction