missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Human IL-17/IL-17A (Research Grade Secukinumab Biosimilar) Antibody, Novus Biologicals™

Human Monoclonal Antibody
248.00 € - 572.00 €
Specifications
| Antigen | IL-17/IL-17A |
|---|---|
| Clone | Hu71 |
| Applications | ELISA |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18784344
![]() |
Novus Biologicals™
MAB9492-SP |
25 μg |
248.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18764314
![]() |
Novus Biologicals™
MAB9492-100 |
100 μg |
572.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-17/IL-17A Monoclonal antibody specifically detects IL-17/IL-17A in Human samples. It is validated for ELISASpecifications
| IL-17/IL-17A | |
| ELISA | |
| Unconjugated | |
| Human | |
| Human | |
| CTLA8cytotoxic T-lymphocyte-associated serine esterase 8, Cytotoxic T-lymphocyte-associated antigen 8, IL-17Acytotoxic T-lymphocyte-associated protein 8, IL-17CTLA-8, IL17interleukin-17A, interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8), interleukin 17A | |
| Human IL-17/IL-17A | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| Hu71 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| 3605 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze/thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 6 months, -20°C to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title