missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-17C Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF1234-SP
This item is not returnable.
View return policy
Description
IL-17C Polyclonal specifically detects IL-17C in Human samples. It is validated for Western Blot.
Specifications
| IL-17C | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CX2, Cytokine CX2, IL17C, IL-17CMGC126884, interleukin 17C, interleukin-17C, MGC138401 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 27189 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| Q9P0M4 | |
| IL17C | |
| E. coli-derived recombinant human IL-17C His19-Val197 Accession # Q9P0M4 | |
| 25 μg | |
| Primary | |
| Detects human IL-17C in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with recombinant human (rh) IL-17D and rhIL-17E is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction