Learn More
Human IL-17F Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  R&D Systems MAB13353-SP
 View more versions of this product
Product Code. 18749617
  • 238.00 € / 25µg
Description
IL-17F Monoclonal specifically detects IL-17F in Human samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IL-17F
Monoclonal
Unconjugated
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS.
Cytokine ML-1, IL17F, IL-17Finterleukin-17F, interleukin 17F
Mouse myeloma cell line NS0-derived recombinant human IL-17F Gly21-Thr153 Accession # Q96PD4
25 μg
112744
Reconstitute at 0.5mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG2b
ELISA
775602R
ELISA
Q96PD4
Mouse
Protein A or G purified from cell culture supernatant
Primary
Detects human IL-17F in direct ELISAs.
Human
Supernatant