missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-17F Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB13353-100
This item is not returnable.
View return policy
Description
IL-17F Monoclonal specifically detects IL-17F in Human samples. It is validated for ELISA.
Specifications
| IL-17F | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
| Cytokine ML-1, IL17F, IL-17Finterleukin-17F, interleukin 17F | |
| Mouse myeloma cell line NS0-derived recombinant human IL-17F Gly21-Thr153 Accession # Q96PD4 | |
| 100 μg | |
| 112744 | |
| Reconstitute at 0.5mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| ELISA | |
| 775602R | |
| ELISA | |
| Q96PD4 | |
| Mouse | |
| Protein A or G purified from cell culture supernatant | |
| Primary | |
| Detects human IL-17F in direct ELISAs. | |
| Human | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction