missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-18/IL-1F4 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems MAB9124
This item is not returnable.
View return policy
Description
IL-18/IL-1F4 Monoclonal specifically detects IL-18/IL-1F4 in Human samples. It is validated for Neutralization.
Specifications
| IL-18/IL-1F4 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Iboctadekin, IFN-gamma-inducing factor, IGIF, IGIFIL-1 gamma, IL18, IL-18MGC12320, IL-1F4, IL1F4iboctadekin, IL-1g, Interferon gamma-inducing factor, interleukin 18 (interferon-gamma-inducing factor), Interleukin-1 gamma, interleukin-18 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 3606 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG |
| Neutralization | |
| 1072F | |
| Neutralization 0.05-0.3 ug/mL | |
| Q14116 | |
| IL18 | |
| E. coli-derived recombinant human IL-18/IL-1F4 Tyr37-Asp197 Accession # Q14116 | |
| 100 μg | |
| Primary | |
| Detects human IL-18/IL-1F4 in direct ELISAs. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction