missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-18 R alpha/IL-1 R5 PE-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 6 publications
279.00 € - 502.00 €
Specifications
| Antigen | IL-18R alpha/IL-1 R5 |
|---|---|
| Clone | 70625 |
| Dilution | Flow Cytometry 10 uL/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18793418
![]() |
R&D Systems
FAB840P-025 |
25 Tests |
279.00 €
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18778016
![]() |
R&D Systems
FAB840P-100 |
100 Tests |
502.00 €
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-18R alpha/IL-1 R5 Monoclonal specifically detects IL-18R alpha/IL-1 R5 in Human samples. It is validated for Flow Cytometry.Specifications
| IL-18R alpha/IL-1 R5 | |
| Flow Cytometry 10 uL/10^6 cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| 8809 | |
| Mouse myeloma cell line NS0-derived recombinant human IL-18 R alpha/IL-1 R5 Glu20-Arg329 Accession # Q13478 | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
| 70625 | |
| Flow Cytometry | |
| R-PE | |
| Mouse | |
| Human | |
| Q13478 | |
| IL18R1 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Detects human IL-18 R alpha/IL-1 R5 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) IL-1 RI, rhIL-1 RII, rhIL-1 R3, rhIL-1 R4, rhIL-1 R6, rhIL-1 R7, rhIL-1 R8, rhIL-1 R9, rhSIGIRR or recombinant mouse IL-18 R alpha is observed. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title