missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-1Rrp2/IL-1R6 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB8721-100
This item is not returnable.
View return policy
Description
IL-1Rrp2/IL-1R6 Monoclonal specifically detects IL-1Rrp2/IL-1R6 in Human samples. It is validated for Neutralization.
Specifications
| IL-1Rrp2/IL-1R6 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| IL-1 R6, IL-1 Rrp2, IL1RL2, IL-1RL2, IL1Rrp2, IL-1Rrp2, IL1RRP2IL1R-rp2Interleukin-1 receptor-related protein 2, interleukin 1 receptor-like 2, interleukin-1 receptor-like 2 | |
| Mouse myeloma cell line NS0-derived recombinant human IL-1 Rrp2/IL-1 R6 Asp20-Tyr337 Accession # Q9HB29 | |
| 100 μg | |
| Primary | |
| Detects human IL-1 Rrp2/IL-1 R6 in direct ELISAs. | |
| Human | |
| Purified |
| Neutralization | |
| 116004 | |
| Neutralization 0.25-1.5 ug/mL | |
| Q9HB29 | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 8808 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction