missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-1Rrp2/IL-1R6 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Brand: R&D Systems AF872
This item is not returnable.
View return policy
Description
IL-1Rrp2/IL-1R6 Polyclonal specifically detects IL-1Rrp2/IL-1R6 in Human samples. It is validated for Western Blot.
Specifications
| IL-1Rrp2/IL-1R6 | |
| Polyclonal | |
| Unconjugated | |
| Q9HB29 | |
| IL1RL2 | |
| Mouse myeloma cell line NS0-derived recombinant human IL-1 Rrp2/IL-1 R6 Asp20-Tyr337 Accession # Q9HB29 | |
| 100 μg | |
| Primary | |
| Detects human IL-1 Rrp2/IL-1 R6 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 15% cross-reactivity with recombinant rat IL-1 R6 is observed and less than 1% cross-reactivity with recombinant human (rh) IL-1 RI and rhIL-1 RII is observed. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| IL-1 R6, IL-1 Rrp2, IL1RL2, IL-1RL2, IL1Rrp2, IL-1Rrp2, IL1RRP2IL1R-rp2Interleukin-1 receptor-related protein 2, interleukin 1 receptor-like 2, interleukin-1 receptor-like 2 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 8808 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction