Learn More
Human IL-31RA Antibody, R&D Systems™
Rat Monoclonal Antibody
Brand:  R&D Systems MAB2769-100
 View more versions of this product
Product Code. 18110041
  • 378.00 € / 100µg
Description
IL-31RA Monoclonal specifically detects IL-31RA in Human samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IL-31RA
Monoclonal
Unconjugated
Lyophilized from a 0.2um filtered solution in Tris, NaCl with Trehalose. *Small pack size (SP) is supplied as a 0.2um filtered solution in PBS.
class I cytokine receptor, CRL3, CRL3Glmr, Cytokine receptor-like 3, GLMR, GLM-R, GLM-RMGC125346, GP130 like receptor, Gp130-like monocyte receptor, Gp130-like receptor, hGLM-R, IL-31 RA, IL-31 receptor subunit alpha, IL-31R subunit alpha, IL31RA, IL-31RAGPLCRL, IL-31R-alpha, interleukin 31 receptor A, interleukin-31 receptor subunit alpha, PRO21384, soluble type I cytokine receptor CRL3, ZcytoR17
NS0-derived rhIL-31 RA extracellular domain aa 20-516 Accession No. NP_620586
100 ug
133396
Reconstitute at 0.5mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG1
Flow Cytometry
313308
Flow Cytometry 0.25ug/106 cells
NP_620586
Rat
Protein A or G purified from hybridoma culture supernatant
Primary
Detects human IL-31 RA in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse IL-31 RA is observed.
Human
Supernatant