Learn More
Human IL-33 Antibody, R&D Systems™
GREENER_CHOICE
Goat Monoclonal Antibody
Brand:  R&D Systems MAB36254-100
 View more versions of this product
Product Code. 18740599
  • 451.00 € / 100µg
Description
IL-33 Monoclonal specifically detects IL-33 in Human samples. It is validated for Western Blot, Immunohistochemistry, Neutralization.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IL-33
Monoclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
C9orf26, C9orf26chromosome 9 open reading frame 26 (NF-HEV), DKFZp586H0523, DVS27, DVS27-related protein, IL1F11, IL-1F11, IL33, IL-33, interleukin 33, Interleukin-1 family member 11, interleukin-33, NFHEV, NF-HEV, NF-HEVNFEHEV, Nuclear factor from high endothelial venules, RP11-575C20.2
Goat
Protein A or G purified from cell culture supernatant
RUO
90865
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG
Western Blot, Immunohistochemistry, Neutralization
40015D
Western Blot 1 ug/mL, Immunohistochemistry 0.3-25 ug/mL, Neutralization 0.75-3 ug/mL
O95760
IL33
E. coli-derived recombinant human IL-33 Ser112-Thr270 Accession # O95760
100 μg
Primary
Detects human IL-33 in direct ELISAs.
Human
Purified