missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-36 alpha/IL-1F6 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems MAB10781-SP
This item is not returnable.
View return policy
Description
IL-36 alpha/IL-1F6 Monoclonal specifically detects IL-36 alpha/IL-1F6 in Human samples. It is validated for Western Blot, Neutralization.
Specifications
| IL-36 alpha/IL-1F6 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| FIL1, FIL1 epsilon, FIL1(EPSILON), FIL1E, IL-1 epsilon, IL1(EPSILON), IL1E, IL-1E, IL1F6, IL-1F6, IL-1F6 (FIL-1-epsilon), IL36 alpha, IL36A, interleukin 1 family, member 6 (epsilon), interleukin 1, epsilon, Interleukin 36, Alpha, Interleukin-1 epsilon, interleukin-1 family member 6, Interleukin-36 Alpha | |
| E. coli-derived recombinant human IL-36 alpha/IL-1F6 Lys6-Phe158 Accession # Q9UHA7 | |
| 25 μg | |
| Primary | |
| Detects human IL-36 alpha/IL-1F6 in direct ELISAs and Western blots. | |
| Human | |
| Purified |
| Western Blot, Neutralization | |
| 1087L | |
| Western Blot 1 ug/mL, Neutralization 0.2-1.2 ug/mL | |
| Q9UHA7 | |
| Rabbit | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 27179 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction