missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-4 Alexa Fluor™ 750-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems IC11474S-100UG
This item is not returnable.
View return policy
Description
IL-4 Monoclonal antibody specifically detects IL-4 in Human samples. It is validated for Flow Cytometry (Intracellular Staining)
Specifications
| IL-4 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| P05112 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3565 | |
| Protect from light. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months, 2 to 8 °C under sterile conditions after opening. | |
| IgG2a |
| Flow Cytometry (Intracellular Staining) | |
| 1067629 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| B cell growth factor 1, B-cell stimulatory factor 1, BCGF1, BCGF-1, binetrakin, BSF1, BSF-1, IL-4B_cell stimulatory factor 1, interleukin 4, interleukin-4, Lymphocyte stimulatory factor 1, MGC79402, Pitrakinra | |
| Chinese Hamster Ovary cell line, CHO-derived human IL-4, His25-Ser153, Accession # P05112 | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction