missing translation for 'onlineSavingsMsg'
Learn More

Human IL-4 Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody

248.00 € - 549.00 €

Specifications

Antigen IL-4
Clone 1067629
Dilution Intracellular Staining by Flow Cytometry 0.25 μg/10^6 cells
Applications Flow Cytometry (Intracellular Staining)
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18784554
View Documents
R&D Systems
MAB11474-100
100 μg
549.00 €
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18764304
GREENER_CHOICE
View Documents
R&D Systems
MAB11474-SP
25 μg
248.00 €
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

IL-4 Monoclonal antibody specifically detects IL-4 in Human samples. It is validated for Flow Cytometry (Intracellular Staining)
Specifications

Specifications

IL-4
Intracellular Staining by Flow Cytometry 0.25 μg/10^6 cells
Monoclonal
Purified
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS.
B cell growth factor 1, BCDF, B-cell stimulatory factor 1, BCGF1, BCGF-1, binetrakin, BSF1, BSF-1, IL4, IL-4B_cell stimulatory factor 1, IL4E12, interleukin 4, interleukin-4, Lymphocyte stimulatory factor 1, MGC79402, Pitrakinra
Chinese Hamster Ovary cell line, CHO-derived human IL-4, His25-Ser153, Accession # P05112
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
1067629
Flow Cytometry (Intracellular Staining)
Unconjugated
Mouse
Human
P05112
3565
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.