missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-4R alpha Alexa Fluor 700-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
288.00 € - 564.00 €
Specifications
| Antigen | IL-4R alpha |
|---|---|
| Clone | 25463 |
| Dilution | Flow Cytometry 5 uL/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18734409
![]() |
R&D Systems
FAB230N-025 |
25 Tests |
288.00 €
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18759047
![]() |
R&D Systems
FAB230N-100 |
100 Tests |
564.00 €
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-4R alpha Monoclonal specifically detects IL-4R alpha in Human samples. It is validated for Flow Cytometry.Specifications
| IL-4R alpha | |
| Flow Cytometry 5 uL/10^6 cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| CD124, CD124 antigen, IL-4 R alpha, IL-4 receptor subunit alpha, IL4R, IL4R alpha, IL-4R subunit alpha, IL-4RA, IL4RACD124, IL-4R-alpha, interleukin 4 receptor, interleukin-4 receptor alpha chain, interleukin-4 receptor subunit alpha | |
| IL4R | |
| IgG2a | |
| Protein A or G purified from ascites | |
| Detects human IL-4 R alpha in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) IL-9 R, recombinant mouse IL-4 R, rhIL-5 R alpha , rhIL-5 R beta , rhIL-13 R alpha 1, or rhIL-13 R alpha 2 is observed. |
| 25463 | |
| Flow Cytometry | |
| Alexa Fluor 700 | |
| Mouse | |
| Human | |
| P24394 | |
| 3566 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IL-4 R Gly24-His232 Accession # P24394 | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title