missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-4R alpha APC-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 5 publications
Brand: R&D Systems FAB230A-025
This item is not returnable.
View return policy
Description
IL-4R alpha Monoclonal specifically detects IL-4R alpha in Human samples. It is validated for Flow Cytometry.
Specifications
| IL-4R alpha | |
| Monoclonal | |
| APC | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| CD124, CD124 antigen, IL-4 R alpha, IL-4 receptor subunit alpha, IL4R, IL4R alpha, IL-4R subunit alpha, IL-4RA, IL4RACD124, IL-4R-alpha, interleukin 4 receptor, interleukin-4 receptor alpha chain, interleukin-4 receptor subunit alpha | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 3566 | |
| Human | |
| Purified |
| Flow Cytometry | |
| 25463 | |
| Flow Cytometry 10 uL/10^6 cells | |
| P24394 | |
| IL4R | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IL-4 R Gly24-His232 Accession # P24394 | |
| 25 Tests | |
| Primary | |
| Detects human IL-4 R alpha in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) IL-9 R, recombinant mouse IL-4 R, rhIL-5 R alpha , rhIL-5 R beta , rhIL-13 R alpha 1, or rhIL-13 R alpha 2 is observed. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction