missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-8/CXCL8 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 96 publications
Brand: R&D Systems MAB208-100
This item is not returnable.
View return policy
Description
CXCL8/IL-8 Monoclonal specifically detects CXCL8/IL-8 in Human samples. It is validated for Western Blot, Simple Western, Neutralization, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready, ELISA Capture (Matched Antibody Pair).
Specifications
| CXCL8/IL-8 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 3-10C, AMCF-I, C-X-C motif chemokine 8, CXCL8, CXCL8SCYB8, Emoctakin, GCP1, GCP-1TSG-1, IL8, interleukin 8, K60, LAI, LECT, LUCT, LYNAP, MDNCF, MDNCFb-ENAP, member 8, MONAP, MONAPGCP1, NAF, NAP1, NAP-1NAP1, NCF, Neutrophil-activating protein 1, Protein 3-10C, T cell chemotactic factor, T-cell chemotactic factor, TCF, TSG1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3576 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot, Neutralization, Flow Cytometry, Immunocytochemistry, CyTOF, ELISA | |
| 6217 | |
| Western Blot 3 ug/mL, Simple Western 50 ug/mL, Neutralization 0.08-0.4 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL, CyTOF-ready, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| P10145 | |
| IL8 | |
| E. coli-derived recombinant human IL-8/CXCL8, Ser28-Ser99, Accession # P10145 | |
| 100 μg | |
| Primary | |
| Detects human IL-8/CXCL8 in ELISAs and Western blots. In Western blots, 100% cross-reactivity with recombinant porcine IL-8/CXCL8 and no cross-reactivity with recombinant rat CXCL3/CINC-2 alpha is observed. | |
| Human | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction