missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Human Integrin beta 1/CD29 - Ready-To-Use ELISA Kit (Colorimetric)

Product Code. 30238837 Shop All Bio Techne Products
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 30238837

Brand: Novus Biologicals™ NBP339962

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Colorimetric ELISA Kit

TRUSTED_SUSTAINABILITY

Specifications

Assay Range 0.781 - 50 ng/mL (example only; lot dependent)
Assay Sensitivity 0.27 ng/mL (example only; lot dependent)
Detection Method Colorimetric
Immunoassay Kit Format Pre-coated 96T Strip Plate
Product Type ELISA Kit (Colorimetric)
Sample Type Tissue Homogenates, Cell Lysates, Cell Culture Supernatant, Other Biological Fluids
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 3688
Gene Symbol ITGB1
Quantity 1 Kit
Research Discipline Cancer, Cell Biology, Cellular Markers, Cytokine Research, Immunology, Mesenchymal Stem Cell Markers, Neuronal Cell Markers, Phospho Specific, Signal Transduction, Stem Cell Markers
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym CD29, CD29 antigen, Fibronectin receptor subunit beta, FNRBVLAB, GPIIA, integrin beta-1, integrin VLA-4 beta subunit, integrin, beta 1 (fibronectin receptor, beta polypeptide, antigen CD29 includesMDF2, MSK12), MDF2, MSK12, very late activation protein, beta polypeptide, VLA-4 subunit beta, VLA-BETA
Target Human
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.