missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human KA2/GRIK5 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Marca: R&D Systems MAB113341-100
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
KA2/GRIK5/Glutamate Receptor KA2 Monoclonal antibody specifically detects KA2/GRIK5/Glutamate Receptor KA2 in Human samples. It is validated for Immunohistochemistry
Especificaciones
| KA2/GRIK5/Glutamate Receptor KA2 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| EAA2, Excitatory amino acid receptor 2, GluK5, glutamate receptor KA2, Glutamate receptor KA-2, glutamate receptor, ionotropic kainate 5, glutamate receptor, ionotropic, kainate 5, GRIK2, GRIK5, KA2GRIK2 | |
| Synthetic peptide sequence, Accession # Q16478 | |
| 100 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 6 months, -20°C to -70°C under sterile conditions after reconstitution. | |
| IgG |
| Immunohistochemistry | |
| 2916E | |
| Immunohistochemistry 5-15 ug/mL | |
| Q16478 | |
| Rabbit | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2901 | |
| Human | |
| Purified |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido