missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Kallikrein 14 Antibody, R&D Systems™

Description
Kallikrein 14 Monoclonal specifically detects Kallikrein 14 in Human samples. It is validated for Immunoprecipitation, Neutralization.
Specifications
Specifications
| Antigen | Kallikrein 14 |
| Applications | Immunoprecipitation, Neutralization |
| Classification | Monoclonal |
| Clone | 337903 |
| Concentration | LYOPH |
| Conjugate | Unconjugated |
| Dilution | Immunoprecipitation 25 ug/mL, Neutralization 0.07 ug/mL |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative |
| Gene Accession No. | AAD50773 |
| Gene Alias | EC 3.4.21, EC 3.4.21.-, hK14, kallikrein 14, Kallikrein-like protein 6, kallikrein-related peptidase 14, KLK14, KLKL6, KLK-L6kallikrein-14 |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?