missing translation for 'onlineSavingsMsg'
Learn More

Human Kallikrein 14 Antibody, R&D Systems™
GREENER_CHOICE

Product Code. p-7065205
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18777169 25 μg 25µg
18143361 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18777169 Supplier R&D Systems Supplier No. MAB2626SP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Kallikrein 14 Monoclonal specifically detects Kallikrein 14 in Human samples. It is validated for Immunoprecipitation, Neutralization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Kallikrein 14
Applications Immunoprecipitation, Neutralization
Classification Monoclonal
Clone 337903
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunoprecipitation 25 ug/mL, Neutralization 0.07 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. AAD50773
Gene Alias EC 3.4.21, EC 3.4.21.-, hK14, kallikrein 14, Kallikrein-like protein 6, kallikrein-related peptidase 14, KLK14, KLKL6, KLK-L6kallikrein-14
Gene Symbols KLK14
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Kallikrein 14 Gln19-Met248 Accession # AAD50773
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 43847
Test Specificity Detects human Kallikrein 14 in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.