missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Human Kidney Whole Tissue Lysate (Adult Whole Primary and Metastatic Tumor Paired)

Product Code. 18293160 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 Set
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18293160 1 Set 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18293160 Supplier Novus Biologicals™ Supplier No. NBL126529
  • ONLINE EXCLUSIVE PRICE
    1965.00 €
    1854.30 € / 1 sets
    Save 110.70 €
    5% Off
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy
The Human Kidney Whole Tissue Lysate (Adult Whole Primary and Metastatic Tumor Paired) is derived from human.
TRUSTED_SUSTAINABILITY

Specifications

Tissue Digestive/Waste
Quantity 1 Set
Storage Buffer HEPES (pH 7.9), MgCl2, KCl, EDTA, Sucrose, Glycerol, Sodium deoxycholate, NP-40, and a cocktail of protease inhibitors
Test Specificity Matched Pair - Total Protein - Human Primary Tumor and Metastatic Tumor Tissue (PM): Kidney
Cellular Fraction Whole
Tissue Condition Primary and Metastatic Tumor Paired
Cross Reactivity Human
Host Species Human
Product Type Human Kidney Whole Tissue Lysate (Adult Primary and Metastatic Tumor Paired)
Content And Storage -80°C. Avoid freeze-thaw cycles.
Life Stage Adult
Protein State Native
Show More Show Less

For Research Use Only

Product Title
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.