missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human KIR2DL1/KIR2DS5 Fluorescein-conjugated Antibody, R&D Systems™
Description
KIR2DL1/KIR2DS5 Monoclonal specifically detects KIR2DL1/KIR2DS5 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
| Antigen | KIR2DL1/KIR2DS5 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 143211 |
| Conjugate | Fluorescein |
| Dilution | Flow Cytometry 10 uL/10^6 cells |
| Formulation | Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide |
| Gene Accession No. | P43626 |
| Gene Alias | CD158a, CD158g, NKAT9 |
| Gene Symbols | KIR2DL1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?