missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Kynureninase Antibody, R&D Systems™

Mouse Monoclonal Antibody
210.00 € - 536.00 €
Specifications
| Antigen | Kynureninase |
|---|---|
| Clone | 589731R |
| Dilution | Western Blot 2 ug/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18711297
![]() |
R&D Systems
MAB4887R-SP |
25 μg |
210.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18742287
![]() |
R&D Systems
MAB4887R-100 |
100 μg |
536.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Kynureninase Monoclonal specifically detects Kynureninase in Human samples. It is validated for Western Blot.Specifications
| Kynureninase | |
| Western Blot 2 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS. | |
| EC 3.7.1.3, KYNU, kynureninase, kynureninase (L-kynurenine hydrolase), L-kynurenine hydrolase | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Kynureninase, Met1-Asn465, Accession # Q16719 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Kynureninase in direct ELISAs. |
| 589731R | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q16719 | |
| 8942 | |
| IgG1 | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title