missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Lactate Dehydrogenase B/LDHB Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems MAB9205-SP
This item is not returnable.
View return policy
Description
Lactate Dehydrogenase B Monoclonal specifically detects Lactate Dehydrogenase B in Human samples. It is validated for Western Blot, Immunohistochemistry, Intracellular Staining by Flow Cytometry, Immunocytochemistry, Knockout Validated.
Specifications
| Lactate Dehydrogenase B | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| EC 1.1.1, EC 1.1.1.27, Epididymis Secretory Protein Li 281, HEL-S-281, lactate dehydrogenase B, LDH heart subunit, LDHB, LDH-B, LDHBD, LDH-H, L-lactate dehydrogenase B chain, Renal Carcinoma Antigen NY-REN-46, TRG-5 | |
| E. coli-derived recombinant human LDHB Met1-Val93 Accession # P07195 | |
| 25 μg | |
| Primary | |
| Detects human LDHB in direct ELISAs and Western blots. | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Flow Cytometry, Immunocytochemistry, KnockDown | |
| 2057D | |
| Western Blot 0.05 ug/mL, Immunohistochemistry 1-25 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 3-25 ug/mL, Knockout Validated | |
| P07195 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 3945 | |
| Reconstitue at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction