missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human LAG-3 Antibody, Novus Biologicals™

Mouse Monoclonal Antibody
221.00 € - 612.00 €
Specifications
| Antigen | LAG-3 |
|---|---|
| Clone | 1058221 |
| Dilution | Immunohistochemistry 5-25 ug/mL, Immunocytochemistry 8-25 ug/mL |
| Applications | Immunohistochemistry, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18726055
![]() |
R&D Systems
MAB231971-100 |
100 μg |
612.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18765735
![]() |
R&D Systems
MAB231971-SP |
25 μg |
221.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LAG-3 Monoclonal antibody specifically detects LAG-3 in Human samples. It is validated for Immunohistochemistry, ImmunocytochemistrySpecifications
| LAG-3 | |
| Immunohistochemistry 5-25 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| CD223, CD223 antigen, LAG3, LAG-3, lymphocyte activating 3, lymphocyte activation gene 3 protein, lymphocyte-activation gene 3, Secreted lymphocyte activation gene 3 protein, sLAG-3 | |
| Human Lag-3 Peptide,, Accession # P18627 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 1058221 | |
| Immunohistochemistry, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| P18627 | |
| 3902 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title