missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human LBP Antibody, R&D Systems™

Mouse Monoclonal Antibody
209.00 € - 397.00 €
Specifications
| Antigen | LBP |
|---|---|
| Clone | 127420 |
| Dilution | ELISA |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18780539
![]() |
R&D Systems
MAB8701-SP |
25 μg |
209.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18797399
![]() |
R&D Systems
MAB8701-100 |
100 μg |
397.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LBP Monoclonal specifically detects LBP in Human samples. It is validated for ELISA.Specifications
| LBP | |
| ELISA | |
| Monoclonal | |
| Supernatant | |
| Human | |
| AAD21962 | |
| 3929 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 127420 | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
| lipopolysaccharide binding protein, lipopolysaccharide-binding protein, LPS-binding protein, MGC22233 | |
| Mouse myeloma cell line NS0-derived recombinant human LBP Ala26-Val481 Accession # AAD21962 | |
| Primary | |
| Reconstitute at 0.5mg/mL in sterile PBS. | |
| Detects human LBP in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title