missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Leptin R Antibody, R&D Systems™

Mouse Monoclonal Antibody
238.00 € - 580.00 €
Specifications
| Antigen | Leptin R |
|---|---|
| Clone | 52263R |
| Dilution | Western Blot 1 ug/mL, Immunohistochemistry 0.5-25 ug/mL |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18732177
![]() |
R&D Systems
MAB867R-SP |
25 μg |
238.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18731207
![]() |
R&D Systems
MAB867R-100 |
100 μg |
580.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Leptin R Monoclonal specifically detects Leptin R in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
| Leptin R | |
| Western Blot 1 ug/mL, Immunohistochemistry 0.5-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| B219, CD295, CD295 antigen, DB, DKFZp686B1731, huB219, LEPR, LEP-R, leptin receptor, LeptinR, OB R, OB receptor, OB-R, OBRCD295 | |
| Mouse myeloma cell line NS0-derived recombinant human Leptin R, Phe22-Asp839, Accession # P48357 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 52263R | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| P48357 | |
| 3953 | |
| IgG2b | |
| Protein A or G purified from ascites | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title