Learn More
Human Leptin R MAb (Clone 52209), R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  R&D Systems MAB86712-SP
 View more versions of this product
Product Code. 18731827
  • 203.00 € / 25µg
Description
Leptin R Monoclonal specifically detects Leptin R in Human samples. It is validated for Neutralization.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Leptin R
Monoclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
B219, CD295, CD295 antigen, DB, DKFZp686B1731, huB219, LEPR, LEP-R, leptin receptor, LeptinR, OB R, OB receptor, OB-R, OBRCD295
Mouse myeloma cell line NS0-derived recombinant human Leptin R, Phe22-Asp839, Accession # P48357
25 μg
3953.0
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
IgG2b
Neutralization
52209
Neutralization 0.2-1 ug/mL
P48357
Mouse
Protein A or G purified from hybridoma culture supernatant
Primary
Detects human Leptin R in direct ELISAs.
Human
Supernatant