missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human LILRA1/LILRB1 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB30851
This item is not returnable.
View return policy
Description
LILRA1/LILRB1 Monoclonal specifically detects LILRA1/LILRB1 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.
Specifications
| LILRA1/LILRB1 | |
| Monoclonal | |
| LYOPH | |
| Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| O75019 | |
| LILRA1 | |
| Mouse myeloma cell line NS0-derived recombinant human LILRA1 Pro17-Asn461 Accession # O75019 | |
| 100 μg | |
| Primary | |
| Detects human LILRA1 in direct ELISAs. In direct ELISAs, 100% cross-reactivity with recombinant human (rh) LILRB1 is observed and no cross-reactivity rhILT3, 4, 5, 6, rhLIR-7 or -8 is observed. | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 586326 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD85I, CD85j, ILT2, LIR1, LIR6, LIR-6, MIR7 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 11024 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction