missing translation for 'onlineSavingsMsg'
Learn More

Human LILRB2/CD85d/ILT4 Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18771277
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18771277 25 μg 25µg
18704527 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18771277 Supplier R&D Systems Supplier No. MAB2078RSP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ILT4/CD85d/LILRB2 Monoclonal specifically detects ILT4/CD85d/LILRB2 in Human samples. It is validated for Western Blot, Flow Cytometry, Neutralization, CyTOF-ready.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ILT4/CD85d/LILRB2
Applications Western Blot, Flow Cytometry, Neutralization, CyTOF
Classification Monoclonal
Clone 287219R
Conjugate Unconjugated
Dilution Western Blot 1 μg/mL, Flow Cytometry 0.25 μg/106 cells, Neutralization 0.15-0.9 μg/mL, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. ACT64556
Gene Alias CD85 antigen-like family member D, CD85d, CD85d antigen, Ig-like transcript 4, ILT4, ILT-4, ILT4CD85d, Immunoglobulin-like transcript 4, Leukocyte immunoglobulin-like receptor 2, leukocyte immunoglobulin-like receptor subfamily B member 2, leukocyte immunoglobulin-like receptor subfamily B member 2 soluble isoform, leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), LIR2, LIR2CD85D, LIR-2subfamily A (with TM domain), member 6, member 2, MIR10, MIR10LILRA6, Monocyte/macrophage immunoglobulin-like receptor 10
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human LILRB2/CD85d/ILT4, Gly24-His458, Accession # ACT64556
Purification Method Protein A or G purified from cell culture supernatant
Quantity 25 μg
Primary or Secondary Primary
Gene ID (Entrez) 10288
Test Specificity Detects human LILRB2/CD85d/ILT4 in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.