missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human LILRB5/CD85c/LIR-8 Antibody, R&D Systems™

Mouse Monoclonal Antibody
228.00 € - 582.00 €
Specifications
| Antigen | LIR-8/CD85c/LILRB5 |
|---|---|
| Clone | 1058033 |
| Dilution | Immunocytochemistry 8-25 ug/mL |
| Applications | Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18702827
![]() |
R&D Systems
MAB30651-SP |
25 μg |
228.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18703577
![]() |
R&D Systems
MAB30651-100 |
100 μg |
582.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LIR-8/CD85c/LILRB5 Monoclonal antibody specifically detects LIR-8/CD85c/LILRB5 in Human samples. It is validated for ImmunocytochemistrySpecifications
| LIR-8/CD85c/LILRB5 | |
| Immunocytochemistry 8-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| CD85 antigen-like family member C, CD85c, CD85c antigen, Leukocyte immunoglobulin-like receptor 8, leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), LIR8, LIR-8, LIR8CD85C, LIR-8leukocyte immunoglobulin-like receptor subfamily B member 5, member 5 | |
| Mouse myeloma cell line, NS0-derived human LILRB5/CD85c/LIR-8, Arg18-His456, Accession # O75023 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 1058033 | |
| Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| O75023 | |
| 10990 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title