missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Human Liver Membrane Tissue Lysate (Adult Membrane Liver Cirrhosis)

Product Code. 18236550 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18236550 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18236550 Supplier Novus Biologicals™ Supplier No. NBL126449
  • ONLINE EXCLUSIVE PRICE
    701.00 €
    662.55 € / 0.1mg
    Save 38.45 €
    5% Off
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy
The Human Liver Membrane Tissue Lysate (Adult Membrane Liver Cirrhosis) is derived from human.
TRUSTED_SUSTAINABILITY

Specifications

Tissue Digestive/Waste
Quantity 0.1 mg
Storage Buffer HEPES (pH 7.9), MgCl2, KCl, EDTA, Sucrose, Glycerol, Sodium deoxycholate, NP-40, and a cocktail of protease inhibitors
Test Specificity Membrane Protein - Liver Cirrhosis: Liver
Cellular Fraction Membrane
Tissue Condition Liver Cirrhosis
Cross Reactivity Human
Host Species Human
Product Type Human, Liver Membrane Tissue Lysate (Adult Liver Cirrhosis)
Content And Storage -80°C. Avoid freeze-thaw cycles.
Life Stage Adult
Protein State Native
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.