missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Human Liver Whole Tissue Lysate (Adult Whole Normal)

Product Code. 18717483 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18717483 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price
Product Code. 18717483 Supplier Novus Biologicals™ Supplier No. NB82059232
  • ONLINE EXCLUSIVE PRICE
    487.00 €
    459.90 € / 1mg
    Save 27.10 €
    5% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price

Specifications

Preparation Method Western Blot Electrophoresis
Quantity 1 mg
Content And Storage Store at -80°C. Avoid freeze/thaw cycles.
Target Liver
Synonym Liver, Liver lysate, liver lysates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.