missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human LRTM1 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems MAB10046-100
This item is not returnable.
View return policy
Description
LRTM1 Monoclonal specifically detects LRTM1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF-ready.
Specifications
| LRTM1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| HT017, leucine-rich repeat and transmembrane domain-containing protein 1, leucine-rich repeats and transmembrane domains 1 | |
| Synthetic peptide containing human LRTM1, Accession # Q9HBL6 | |
| 100 μg | |
| 57408 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF | |
| 2381D | |
| Western Blot 2 μg/mL, Flow Cytometry 0.25 μg/106 cells, Immunohistochemistry 3-25 μg/mL, Immunocytochemistry 3-25 μg/mL, CyTOF-ready | |
| Q9HBL6 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| Primary | |
| Detects human LRTM1 in direct ELISAs. | |
| Human | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction