missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Human Lung Membrane Tissue Lysate (Adult Membrane Tumor)

Product Code. 18295210 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18295210 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18295210 Supplier Novus Biologicals™ Supplier No. NBL126425
  • ONLINE EXCLUSIVE PRICE
    701.00 €
    662.55 € / 0.1mg
    Save 38.45 €
    5% Off
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy
The Human Lung Membrane Tissue Lysate (Adult Membrane Tumor) is derived from human.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Western Blot Electrophoresis
Tissue Respiratory
Quantity 0.1 mg
Storage Buffer HEPES (pH 7.9), MgCl2, KCl, EDTA, Sucrose, Glycerol, Sodium deoxycholate, NP-40, and a cocktail of protease inhibitors
Test Specificity Membrane Protein - Human Tumor Tissue: Lung
Cellular Fraction Membrane
Tissue Condition Tumor
Cross Reactivity Human
Host Species Human
Concentration 2 mg/mL
Product Type Human Lung Membrane Tissue Lysate (Adult Tumor)
Content And Storage -80°C. Avoid freeze-thaw cycles.
Life Stage Adult
Protein State Native
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.