missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Lysosomal Pro-X Carboxypeptidase/PRCP Antibody, R&D Systems™
Rat Monoclonal Antibody
Brand: R&D Systems MAB7164
This item is not returnable.
View return policy
Description
Lysosomal Pro-X Carboxypeptidase/PRCP Monoclonal specifically detects Lysosomal Pro-X Carboxypeptidase/PRCP in Human samples. It is validated for Western Blot.
Specifications
| Lysosomal Pro-X Carboxypeptidase/PRCP | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| P42785 | |
| PRCP | |
| Mouse myeloma cell line NS0-derived recombinant human Lysosomal Pro-X Carboxypeptidase/PRCP Met1-His496 Accession # P42785 | |
| 100 μg | |
| Primary | |
| Detects human Lysosomal Pro-X Carboxypeptidase/PRCP in ELISAs. | |
| Human | |
| Purified |
| Western Blot | |
| 860821 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Angiotensinase C, EC 3.4.16.2, HUMPCP, Lysosomal carboxypeptidase C, lysosomal Pro-X carboxypeptidase, PCPMGC2202, Proline carboxypeptidase, Prolylcarboxypeptidase, prolylcarboxypeptidase (angiotensinase C), prolylcarboxypeptidase isoform 1 preproprotein | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5547 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction